Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSG101 Rabbit anti-Human, Rat, Clone: 4T4W6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | TSG101 |
---|---|
Clone | 4T4W6 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
TSG101 Monoclonal antibody specifically detects TSG101 in Human, Rat samples. It is validated for Western BlotSpecifications
TSG101 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Autophagy, Cancer, Microautophagy | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
7251 | |
IgG | |
Affinity purified |
4T4W6 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
ESCRT-I complex subunit TSG101, TSG10, tumor susceptibility gene 10, tumor susceptibility gene 101, tumor susceptibility gene 101 protein, tumor susceptibility protein, VPS23 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TSG101 (Q99816). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title