Learn More
TSG101 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | TSG101 |
---|---|
Concentration | 267 μg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8656
![]() ![]() |
Proteintech Group Inc
282831AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The TSG101 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TSG101. This antibody recognizes human, mouse antigen. The TSG101 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
TSG101 | |
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
Unconjugated | |
Human, Mouse | |
IgG |
267 μg/mL | |
Polyclonal | |
Rabbit | |
7251 | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.