Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSGA10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571349
Description
TSGA10 Polyclonal Antibody for Western Blot
TSGA10 may play a role in the sperm tail fibrous sheath, a major sperm tail structure.
Specifications
TSGA10 | |
Polyclonal | |
Unconjugated | |
TSGA10 | |
4933432N21Rik; AI843383; cancer/testis antigen 79; CEP4L; Cp431; CT79; Gm217; Mtsga10; testis development protein NYD-SP7; testis specific 10; testis specific, 10; testis-specific gene 10 protein; Tsga10 | |
Rabbit | |
Affinity chromatography | |
RUO | |
80705 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BZW7 | |
TSGA10 | |
synthetic peptide directed towards the N-terminal region of human TSGA10. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction