Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSGA10IP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548194
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548194 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548194 Supplier Thermo Scientific Supplier No. PA548194
Add to Cart
Add to Cart

TSGA10IP Polyclonal Antibody

TSGA10IP Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen TSGA10IP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TSGA10IP
Gene Accession No. Q3SY00
Gene Alias FAM161C; testis specific 10 interacting protein; testis specific, 10 interacting protein; testis-specific protein 10-interacting protein; Tsga10-interacting protein; TSGA10IP
Gene Symbols TSGA10IP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 492-520 amino acids from the C-terminal region of human TSGA10IP.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254187
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.