Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSGA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553601
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553601 Supplier Invitrogen™ Supplier No. PA553601
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TAELIHLNHR YQGKFLNKNP VGPGKYVFDV GCEQHGEYRL TDMERGEEEE EEELVTVVPK WKATQITELA LWTPTL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.

This gene encodes a male meiotic metaphase chromosome-associated acidic protein. This gene is expressed in tissues with motile cilia or flagella, including the trachea, lungs, airway brushings, and testes. Mutations in this gene result in primary ciliary dyskinesia-24. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSGA2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RSPH1
Gene Accession No. Q8WYR4
Gene Alias ABCB2; cancer/testis antigen 79; CT79; h-meichroacidin; Male meiotic metaphase chromosome-associated acidic protein; MCA; Meichroacidin; MGC126568; MGC141927; PSF1; radial spoke head 1 homolog; radial spoke head 1 homolog (Chlamydomonas); RING4; RSP44; Rsph1; RSPH10A; testes specific gene A2 homolog; testis specific A2 homolog; testis specific gene A2; testis-specific gene A2 protein; TSA2; TSGA2; TSGA2 epididymal isoform
Gene Symbols RSPH1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TSGA2. Recombinant protein control fragment (Product #RP-91776).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89765
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.