Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSH b-subunit Monoclonal Antibody (ABM802), Abbomax

Catalog No. 502204182
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4182 Supplier Abbomax Inc Supplier No. 603750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Thyroid Cellular location: Secreted.

TSH b-subunit Monoclonal antibody specifically detects TSH b-subunit in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen TSH b-subunit
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ABM802
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene TSHB
Gene Accession No. P01222
Gene Alias thyroid stimulating hormone beta; thyroid stimulating hormone subunit beta; thyroid stimulating hormone, beta; thyroid stimulating hormone, beta subunit; thyroid-stimulating hormone beta subunit; thyroid-stimulating hormone subunit beta; Thyrotropin alfa; Thyrotropin beta chain; thyrotropin beta subunit; thyrotropin subunit beta; TSH; Tshb; TSH-B; TSH-beta
Host Species Mouse
Immunogen Human TSH B-Chain purified by affinity
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7252
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.