Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSH beta Antibody (154), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062385AF350
Description
TSH beta Monoclonal antibody specifically detects TSH beta in Human samples. It is validated for ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
TSH beta | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry, Immunofluorescence | |
154 | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA | |
Native Human beta-TSH | |
0.5 mL | |
Cancer, Signal Transduction | |
7252 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction