Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSH beta Antibody (SR1242), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32189750UL
Description
TSH beta Monoclonal antibody specifically detects TSH beta in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
TSH beta | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR1242 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA | |
A synthesized peptide derived from human TSH beta (Uniprot #: P01222) | |
50 μg | |
Cancer, Signal Transduction | |
7252 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction