Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSH beta Polyclonal Antibody, Invitrogen™

Catalog No. FEPA5145294
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145294 20 μL
FEPA5145293 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145294 Supplier Invitrogen™ Supplier No. PA5145294
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TSH is a pituitary hormone of 28kDa, which stimulates thyroid growth and production of thyroid hormones. This antibody labels thyrotropic cells of the pituitary.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSH beta
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene TSHB
Gene Accession No. P54828
Gene Alias thyroid stimulating hormone beta; thyroid stimulating hormone subunit beta; thyroid stimulating hormone, beta; thyroid stimulating hormone, beta subunit; thyroid-stimulating hormone beta subunit; thyroid-stimulating hormone subunit beta; Thyrotropin alfa; Thyrotropin beta chain; thyrotropin beta subunit; thyrotropin subunit beta; TSH; Tshb; TSH-B; TSH-beta
Gene Symbols TSHB
Host Species Rabbit
Immunogen Recombinant Dog TSHb protein, Ser20-Ile138 (Accession #P54828).
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 403973
Target Species Canine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.