Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSH Receptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534296
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534296 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534296 Supplier Invitrogen™ Supplier No. PA534296
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset (100%) Bat, Bovine, Cat, Panda, Pig (90%) Sheep, Dog, Horse, Rabbit (85%).

TSH Receptor gene is a membrane protein and a major controller of thyroid cell metabolism. The encoded protein is a receptor for thyrothropin and thyrostimulin, and its activity is mediated by adenylate cyclase. Defects in this gene are a cause of several types of hyperthyroidism. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSH Receptor
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TSHR
Gene Accession No. P16473
Gene Alias AI481368; CHNG1; hTSHR-I; hypothroid; hyt; LGR3; MGC141920; pet; seven transmembrane helix receptor; thyroid stimulating hormone receptor; thyroid stimulating hormone receptor, isoform 2; thyroid-stimulating hormone receptor; thyroid-stimulating hormone receptor variant; thyrotropin receptor; thyrotropin receptor-I, hTSHR-I; TSHR; TSH-R; TSHR; seven transmembrane domain G protein-coupled receptor; TSHRA
Gene Symbols TSHR
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from C-terminus of human TSH Receptor.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7253
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.