Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSHR Antibody (484404), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB65342PECY7
Description
TSHR Monoclonal antibody specifically detects TSHR in Human samples. It is validated for Flow CytometrySpecifications
TSHR | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
484404 | |
Flow Cytometry | |
CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R | |
NS0 mouse myeloma cell line transfected with human TSH R, Accession # P16473 | |
0.1 mL | |
Cell Cycle and Replication, GPCR, Membrane Trafficking and Chaperones, Neuroscience | |
7253 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction