Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSHR Antibody (SPM223), Novus Biologicals™

Mouse Monoclonal Antibody
$280.00 - $536.00
Specifications
Antigen | TSH R |
---|---|
Clone | SPM223 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25321720
![]() |
Novus Biologicals
NBP25321720UG |
20 μg |
Each for $280.00
|
|
|||||
NB253217100
![]() |
Novus Biologicals
NBP253217100UG |
100 μg |
Each for $536.00
|
|
|||||
NB253217200
![]() |
Novus Biologicals
NBP253217200UG |
200 μg | N/A | N/A | N/A | ||||
Description
TSHR Monoclonal specifically detects TSHR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TSH R | |
0.2 mg/mL | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication, GPCR, Membrane Trafficking and Chaperones, Neuroscience | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
7253 | |
Recombinant human TSHR, A-Chain protein | |
Primary | |
Store at 4C. |
SPM223 | |
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R | |
TSHR | |
IgG1 | |
Protein A or G purified | |
62 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title