Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSHR Antibody (SPM223), PE/Cy5.5, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP254569PECY55

Catalog No. N254569PEC5


Only null left
Add to Cart

Description

Description

TSHR Monoclonal specifically detects TSHR in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

TSHR
Monoclonal
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
TSHR
Recombinant human TSHR protein (Uniprot: P16473)
0.1 mL
Primary
Human
IgG1 κ
Flow Cytometry
SPM223
Flow Cytometry
CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R
Mouse
Protein A or G purified
Cell Cycle and Replication, GPCR, Membrane Trafficking and Chaperones, Neuroscience
7253.0
Store at 4°C in the dark. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.