Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSHR Antibody (TSHRB/1404), Novus Biologicals™

Mouse Monoclonal Antibody
$280.00 - $524.00
Specifications
| Antigen | TSH R |
|---|---|
| Clone | TSHRB/1404 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25336320
![]() |
Novus Biologicals
NBP25336320UG |
20 μg |
Each for $280.00
|
|
|||||
NB253363100
![]() |
Novus Biologicals
NBP253363100UG |
100 μg |
Each for $524.00
|
|
|||||
NB253363200
![]() |
Novus Biologicals
NBP253363200UG |
200 μg | N/A | N/A | N/A | ||||
Description
TSHR Monoclonal specifically detects TSHR in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| TSH R | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, GPCR, Membrane Trafficking and Chaperones, Neuroscience | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 7253 | |
| Recombinant human TSHR, B-Chain protein | |
| Primary | |
| Store at 4C. |
| TSHRB/1404 | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R | |
| TSHR | |
| IgG1 | |
| Protein A or G purified | |
| 42 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title