Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSHR Antibody (4C1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$256.00 - $584.50
Specifications
Antigen | TSH R |
---|---|
Clone | 4C1 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1:100-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:1000-1:2000, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry (Paraffin), Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017746
![]() |
Novus Biologicals
NB100657970.025MG |
0.025 mg |
Each for $256.00
|
|
|||||
NB100657971
![]() |
Novus Biologicals
NB100657970.1MG |
0.1 mg |
Each for $584.50
|
|
|||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
TSHR Monoclonal specifically detects TSHR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
TSH R | |
1.0 mg/mL | |
Immunohistochemistry (Paraffin), Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication, GPCR, Membrane Trafficking and Chaperones, Neuroscience | |
PBS (pH 7.4) with 0.09% Sodium Azide | |
CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R | |
TSHR | |
IgG2a | |
Protein A or G purified | |
Recognizes the human thyroid stimulating hormone (TSH) receptor (otherwise known as the thyrotropin receptor) binding to an extracellular domain. |
4C1 | |
Flow Cytometry 1:100-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:1000-1:2000, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P16473 | |
7253 | |
External domain of human TSH receptor. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title