Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSHZ1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582271
Description
Immunogen sequence: YGSPFSESSD QLAHFKGSSS REEKEDPQCP DSVSYPQDSL AQIKAVYANL FSESCWSSLA LDLKKSGSTT STNDASQKES SAPTPTPPTC PVSTTGPTTS TPSTSCSSST SHSSTTSTSS SSGYDWHQAA LAKTLQQTSS YGLLPEPS.
This gene encodes a colon cancer antigen that was defined by serological analysis of recombinant cDNA expression libraries. The encoded protein is a member of the teashirt C2H2-type zinc-finger protein family and may be involved in transcriptional regulation of developmental processes.
Specifications
TSHZ1 | |
Polyclonal | |
Unconjugated | |
TSHZ1 | |
5730407I04Rik; antigen NY-CO-33; CAA; D18Bwg1409e; Kiaa4206; mKIAA4206; Mtsh1; NY-CO-33; SDCCAG33; Serologically defined colon cancer antigen 3 homolog; serologically defined colon cancer antigen 33; Teashirt homolog 1; teashirt zinc finger family member 1; teashirt zinc finger homeobox 1; teashirt1; TSH1; Tshz1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10194 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZSZ6 | |
TSHZ1 | |
Recombinant protein corresponding to Human TSHZ1. Recombinant protein control fragment (Product #RP-89414). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction