Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSHZ2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534343
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534343 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534343 Supplier Invitrogen™ Supplier No. PA534343
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is A-20 cell lysate. PA5-34343 can be used with blocking peptide PEP-1386.

The Teashirt zinc finger homeobox (TSHZ) family comprise a family of evolutionarily conserved transcription factors that, in Drosophila, are active in specific body parts for patterning, but whose function in vertebrates is less clear. In mice, the known three TSHZ proteins are expressed in distinct patterns in the developing and adult brain, suggesting that they play a role in the establishment of regional identity and specification of cell types within the brain. Recent experiments have shown that the expression of TSHZ2 is frequently downregulated in most breast and prostate cancers and its promoter was unmethylated in virtually all cases, suggesting this family of proteins may also be involved in carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSHZ2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TSHZ2
Gene Accession No. Q68FE9, Q9NRE2
Gene Alias 2900073F20Rik; B830045G17; C20orf17; cell growth-inhibiting protein 7; Kiaa4248; mKIAA4248; Mtsh2; Ovarian cancer-related protein 10-2; OVC10-2; Sdccag33l; SDCCAG33-like protein; serologically defined colon cancer antigen 33 like; teashirt 2; teashirt family zinc finger 2; teashirt homolog 2; teashirt homolog 2-like; teashirt zinc finger family member 2; teashirt zinc finger homeobox 2; teashirt2; TSH2; TSHZ2; ZABC2; Zfp218; Zinc finger protein 218; ZNF218
Gene Symbols TSHZ2
Host Species Rabbit
Immunogen A 17 amino acid synthetic peptide near the amino terminus of human TSHZ2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100911757, 128553, 228911
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.