Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Amide 80 HPLC Column, 2 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021724 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
5 cm
10 cm
15 cm
Diameter (Metric):
2 mm
3 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-102-1724 5 cm 2 mm
11-102-1725 10 cm 2 mm
11-102-1726 15 cm 2 mm
11-102-1727 5 cm 3 mm
11-102-1728 10 cm 3 mm
11-102-1729 15 cm 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 11-102-1724 Supplier MilliporeSigma Supelco Supplier No. 80023454
Add to Cart
Add to Cart

TSKgel™ Amide-80 columns are used for the analysis of saccharides, glycans, oligosaccharides, peptides and polar compounds from natural products.

TSKgel™ Amide-80's amide stationary phase provides a unique selectivity under regular normal phase conditions or in the hydrophilic interaction mode of chromatography.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2 mm
Length (Metric) 5 cm
Max. Pressure 150 bar
Particle Size 2 μm
pH 2.0 - 7.5
Surface Area 450 m2/g
Temperature 50°C
USP Type L68
Stationary Phase Carbamoyl
Product Line TSKgel
Matrix Spherical Silica
Phase HILIC
Material Stainless Steel Hardware
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.