Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Amide-80 HPLC Column, 3 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. p-7213970 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
5 cm
10 cm
15 cm
Diameter (Metric):
2 mm
3 mm
4.6 mm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-102-2191 5 cm 3 mm
11-102-2076 5 cm 2 mm
11-102-2192 10 cm 3 mm
11-102-2077 15 cm 2 mm
11-102-2193 15 cm 3 mm
11-102-2078 5 cm 4.6 mm
11-102-2190 10 cm 4.6 mm
11-102-2079 15 cm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 11-102-2191 Supplier MilliporeSigma Supelco Supplier No. 822850
Only null left
Add to Cart
Add to Cart

Target applications include the analysis of saccharides, glycans, oligosaccarides, peptides and polar compounds from natural products.

Unique advantages of the Amide-80 phase for saccharide analysis include novel hydrogen bonding retention mechanism between hydroxyl groups of the sample and the carbamoyl group in the stationary phase. The stationary phase does not react with reducing sugars and can be used at elevated temperatures (4 to 80°C) to prevent peak splitting of carbohydrates that can occur at lower temperatures. In addition, high efficiency 3 micron columns offer shorter analysis time and enhanced sensitivity for LC-MS applications.

Order Info

Shipped in 85% acetonitrile - 15% water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 3 mm
Length (Metric) 5 cm
Max. Pressure 200 bar
Particle Size 3 μm
pH 2.0 - 7.5
Pore Size 100 Å
Solvents 0 to 100% organic solvent
Surface Area 450 m2/g
Temperature 10°C to 50°C
USP Type L68
Stationary Phase Carbamoyl
Product Line TSKgel
Matrix Spherical Silica
Phase HILIC
Material Stainless Steel
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.