Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Amide-80 HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022030 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
5 cm
10 cm
15 cm
25 cm
Diameter (Metric):
1 mm
2 mm
4.6 mm
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-102-2030 5 cm 1 mm
11-102-2031 10 cm 1 mm
11-102-1833 25 cm 4.6 mm
11-102-2056 15 cm 1 mm
11-102-2057 25 cm 1 mm
11-102-2026 5 cm 2 mm
11-102-2027 10 cm 2 mm
11-102-2028 15 cm 2 mm
11-102-2029 25 cm 2 mm
11-102-2010 5 cm 4.6 mm
11-102-2011 10 cm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 11-102-2030 Supplier MilliporeSigma Supelco Supplier No. 820009
Add to Cart
Add to Cart

For the analysis of saccharides, glycans, oligosaccarides, peptides, and polar compounds from natural products.

  • Columns are packed with 3, 5, or 10 micron spherical silica particles covalently bonded with carbamoyl groups
  • Amide stationary phase provides a unique selectivity under regular normal phase conditions, or in the hydrophilic interaction mode of chromatography
  • Target applications include the analysis of saccharides, glycans, oligosaccarides, peptides, and polar compounds from natural products
  • Unique advantages of Amide-80 phase for saccharide analysis include a novel hydrogen bonding retention mechanism between hydroxyl groups of the sample and the carbamoyl group in the stationary phase
  • Stationary phase does not react with reducing sugars
  • Can be used at elevated temperatures (≤ 80°C) to prevent peak splitting of carbohydrates
  • High efficiency, 3 micron columns offer shorter analysis time and enhanced sensitivity for LC-MS applications
  • Shipped in 85% acetonitrile, 15% water
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 1 mm
Length (Metric) 5 cm
Max. Pressure 30 bar
Particle Size 5 μm
pH 2 to 7.5
Pore Size 100 Å
Solvents 0-100% Organic Solvent
Surface Area 450 m2/g
Temperature 10°C to 80°C
USP Type L68
Stationary Phase Carbamoyl
Product Line TSKgel
Shipping Condition Shipped in 85% Acetonitrile, 15% Water
Matrix Carbamoyl Phase/Silica Particle Platform
Material Stainless Steel Hardware
Technique HILIC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.