Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ BioAssist™ Desalting HPLC Column, 15 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022053 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
15 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-2053 15 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2053 Supplier MilliporeSigma Supelco Supplier No. 821463
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for the desalting and buffer exchange of proteins and polynucleotides at semi-preparative scale.

  • Excellent desalting performance
  • Packed with 15 μm polyacrylamide beads in PEEK hardware
  • The novel, hydrophilic, highly cross-linked polyacrylamide beads exhibit superior mechanical strength compared with conventional hydrophilic polyacrylamide beads and cross-linked dextran beads
  • This increase in strength is what allows the use of the small, spherical 15 μm beads
  • Columns can be operated in standard HPLC systems to quickly and efficiently reduce salt and/or buffer concentrations of collected protein or nucleic acid fractions
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 1.5 mL/min
Length (Metric) 15 cm
Max. Pressure 13 bar
Particle Size 15 μm
pH 3 to 12 (≤ 1 Week), 4 to 11 (> 1 Week)
Solvents ≤ 30% Polar Organic Solvent
Temperature 4°C to 30°C
Stationary Phase Polyacrylamide
Product Line TSKgel
Shipping Condition Shipped aq. soln.
Matrix Polyacrylamide Phase/Urea Cross-linked Polyacrylamide
Phase Size Exclusion
Material PEEK
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.