Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ BioAssist™ S HPLC Column, 7 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022025 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-2025 5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2025 Supplier MilliporeSigma Supelco Supplier No. 819686
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Packed with porous, hydrophilic polymer beads that are surface modified with a strong cation exchanger.

  • Packed with spherical, polymer beads (1300 Å)
  • Surface modified with strong sulfopropyl cation exchange groups
  • Large pores allow large biomolecules to freely enter and exit the pores
  • Offers higher binding capacity and better recovery of activity than other competivie materials
  • Standard column dimensions are 4.6 mm ID x 5 cm
  • A 10 mm ID x 10 cm semi-preparative column is available for scale-up
  • Columns are made of PEEK to reduce protein adsorption
  • Salt Concentration: ≤ 2.6 M
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 1 mL/min
Ion Exchange Group Na+
Length (Metric) 5 cm
Max. Pressure 25 bar
Particle Size 7 μm
pH 2 to 12 (≤ 1 Month), 3 to 10 (> 1 Month)
Pore Size 1300 Å
Solvents ≤ 30% Organic Solvent
Temperature 4°C to 60°C
Stationary Phase Sulfonic Acid
Product Line TSKgel
Shipping Condition Shipped in 20% Ethanol in 20 mM Phosphate Buffer, pH 6.5
Matrix Sulfonic Acid Phase/Polymer Particle Platform
Material PEEK Hardware
Technique Cation Exchange
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.