Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ CM-3SW HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021767 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
7.5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1767 7.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1767 Supplier MilliporeSigma Supelco Supplier No. 807162
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel™ CM-3SW columns are packed with 5 micron, high efficiency, spherical silica beads which are surface modified with a weak cation exchanger. They are used for the separation and analysis of proteins, peptides, and other biologically active molecules. They have a larger pore size than the TSKgel™ CM-2SW columns.

  • Stationary Phase: Carboxymethyl (weak cation exchanger)
  • Matrix: Silica particle platform
  • Particle Size: 10 μm
  • Pore Size: 250 Å
  • Dimensions: 7.5 cm x 7.5 mm
  • Operating pH: 2.0 - 7.5
  • Max Pressure: 20 bar
  • Temperature Range: 10°C to 45°C
  • Flow Rate: 1.2 mL/min
  • Molecular Weight Range: 200 - 300,000
  • Compatibility: 0 to 100% organic solvent; ≤1 M salt concentration
  • Counter Ion: Na+
  • Physical Form: Shipped in methanol

Order Info

Shipped in methanol.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7.5 mm
Flow Rate 1.2 mL/min.
Length (Metric) 7.5 cm
Max. Pressure 20 bar
Particle Size 10 μm
pH 2.0 - 7.5
Pore Size 250 Å
Solvents 0 to 100% organic solvent
Temperature 10°C to 45°C
Stationary Phase Carboxymethyl
Product Line TSKgel
Matrix Silica Particle Platform
Phase Cation Exchange
Material Stainless Steel
Technique Cation Exchange
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.