Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ G2000PW HPLC Column, 12 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021744 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
30 cm
60 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1744 30 cm
11-102-1734 60 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-1744 Supplier MilliporeSigma Supelco Supplier No. 805761
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use the TSKgel™ G2000PW HPLC column for analysis of water-soluble polymers with molecular weights up to 2,000 Da.

Order Info

Shipped in water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7.5 mm
Flow Rate 1.2 mL/min.
Length (Metric) 30 cm
Max. Pressure 20 bar
Particle Size 12 μm
pH 2.0 - 12
Pore Size 125 Å
Solvents ≤20% organic solvent
Temperature 80°C
USP Type L25, L38, L39
Stationary Phase Hydroxylated Methacrylate
Product Line TSKgel
Matrix Polymer Particle Platform
Phase Size Exclusion
Material Stainless Steel
Technique SEC HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.