Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ G3000PWXL HPLC Column, 7 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021790 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1790 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1790 Supplier MilliporeSigma Supelco Supplier No. 808021
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use TSKgel™ G3000PWXL columns for the analysis of water-soluble linear polymers with molecular weights up to 50,000 Da.

The smaller particle size of PWXL-type columns provide 1.7 times higher resolution than their PW counterpart, making PWXL-type columns more suitable for analytical purposes.

Order Info

Shipped in water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 1 mL/min.
Length (Metric) 30 cm
Max. Pressure 40 bar
Particle Size 7 μm
pH 2.0 - 12
Pore Size 200 Å
Solvents ≤20% organic solvent
Temperature 80°C
USP Type L37, L38, L39
Stationary Phase Hydroxylated Methacrylate
Product Line TSKgel
Matrix Polymer Particle Platform
Phase Size Exclusion
Material Stainless Steel
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.