Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ G4000PW HPLC Column, 17 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021746 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
30 cm
60 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1746 30 cm
11-102-1736 60 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-1746 Supplier MilliporeSigma Supelco Supplier No. 805763
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel™ G4000PW SEC columns can be used for the water analysis of water-soluble polymers of less than 300,000 Da molecular weight.

  • Separation Technique: Aqueous size exclusion chromatography (GFC)
  • Base Material: Hydroxylated methacrylate polymer
  • Pore Size: 500 Å
  • pH Stability: 2.0 to 12.0
  • Temperature Tolerance: Up to 80°C
  • Max Pressure: 10 bar
  • Mobile Phases: Water or buffer (up to 20% organic solvent)
  • Applications: Separation of water-soluble polymers, oligomers, polysaccharides, nucleic acids, proteins, and peptides

Order Info

Shipped in water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7.5 mm
Flow Rate 1.2 mL/min.
Length (Metric) 30 cm
Max. Pressure 10 bar
Particle Size 17 μm
pH 2.0 - 12
Pore Size 500 Å
Solvents ≤20% organic solvent
Temperature 80°C
USP Type L38, L39
Stationary Phase Hydroxylated Methacrylate
Product Line TSKgel
Matrix Polymer Particle Platform
Phase Size Exclusion
Material Stainless Steel
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.