Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ G4000SW HPLC Column, 17 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021755 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
30 cm
60 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1755 30 cm
11-102-1741 60 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-1755 Supplier MilliporeSigma Supelco Supplier No. 806729
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

TSKgel™ G4000SW SEC columns provide excellent separation of proteins and other biomolecules.

TSKgel™ G4000SW columns are available in 13 μm particle size in glass and stainless steel analytical columns. Particle sizes of 13 and 17 μm are used to pack 30 cm and 60 cm x 21.5 mm I.D. columns, respectively, for semi-preparative purifications.

Order Info

Shipped in 0.05% NaN3 and 0.1 mol/L Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 21.5 mm
Flow Rate 8 mL/min.
Length (Metric) 30 cm
Max. Pressure 10 bar
Particle Size 17 μm
pH 2.5 - 7.5
Pore Size 450 Å
Solvents 0 to 100% aqueous soluble organic solvents
Temperature 10°C to 30°C
USP Type L20, L33, L59
Stationary Phase Diol
Product Line TSKgel
Matrix Silica Particle Platform
Phase Size Exclusion
Material Stainless Steel Hardware
Technique SEC HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.