Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ G4000SWXL HPLC Column, 8 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021809 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1809 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1809 Supplier MilliporeSigma Supelco Supplier No. 808542
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Widely considered the premium columns for analyzing biopolymers by GFC, TSKgel™ SWXL columns feature high pore volume per unit column volume, low sample adsorption and excellent column efficiency, all contributing to unsurpassed sample resolution.

TSKgel™ G4000SWXL columns provide the largest exclusion limit and the widest fractionation range. It is an excellent choice for pegylated proteins or glycosylated biomolecules. The fractionation range is applicable for globular protein samples up to 7 million Da.

Order Info

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 1.2 mL/min.
Length (Metric) 30 cm
Max. Pressure 35 bar
Particle Size 8 μm
pH 2.5 - 7.5
Pore Size 450 Å
Solvents 0 to 100% aqueous soluble organic solvents
Temperature 10°C to 30°C
USP Type L20, L33, L59
Stationary Phase Diol
Product Line TSKgel
Matrix Silica Particle Platform
Phase Size Exclusion
Material Stainless Steel
Technique SEC HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.