Learn More
TSKgel™ GMHXL HPLC Column, 9 μm Particle Size, Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
- Mixed bed, or linear columns are used when a broad range of polymer molecular weights is expected in the sample
- TSKgel GMHxl provides high resolution of polymers with a molecular weight up to several million Da
- THF shipping solvent can be replaced by benzene, chloroform, toluene, and xylene, but not with dichloromethane and dichloroethane
Specifications
Specifications
| Diameter (Metric) | 7.8 mm |
| Flow Rate | 1.2 mL/min |
| Length (Metric) | 30 cm |
| Max. Pressure | 15 bar |
| Particle Size | 9 μm |
| Pore Size | Mixed |
| Temperature | 80°C |
| USP Type | L21 |
| Stationary Phase | Polystyrene-divinylbenzene |
| Product Line | TSKgel |
| Show More |
TSKgel is a registered trademark of Tosoh Corporation
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.