Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ GMPW HPLC Column, 17 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021795 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
30 cm
60 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1795 60 cm
11-102-1794 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-1795 Supplier MilliporeSigma Supelco Supplier No. 808027
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel™ GMPW is a mixed bed scouting column for water-soluble linear polymers. The pore volume can be accessed by polymers ranging in molecular weight from 500 to 8,000,000 Da.

By quickly categorizing the MW profile of an unknown sample, the column enables a fast selection of the best TSKgel™ PW column for routine analysis. Mixed bed columns are also used when a sample has a very broad MW distribution.

Order Info

Shipped in water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7.5 mm
Flow Rate 8 mL/min.
Length (Metric) 60 cm
Max. Pressure 20 bar
Particle Size 17 μm
pH 2.0 - 12
Pore Size <100 Å
Solvents ≤20% organic solvent
Temperature 80°C
USP Type L38, L39
Stationary Phase Hydroxylated Methacrylate
Product Line TSKgel
Matrix Polymer Particle Platform
Phase Size Exclusion
Material Stainless Steel Hardware
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.