Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ GMPWXL HPLC Column, 13 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021793 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1793 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1793 Supplier MilliporeSigma Supelco Supplier No. 808025
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel™ GMPWXL is a mixed bed scouting column for aqueous water-soluble linear polymers. Its pore volume is accessible to polymers ranging from molecular weights of 500 up to 8,000,000 Da.

By quickly categorizing the MW profile of an unknown sample, TSKgel™ GMPWXL allows the selection of the most appropriate TSKgel™ PWXL column for best results in a routine analysis. TSKgel™ GMPWXL is also used for the analysis of samples with broad MW distributions.

Order Info

Shipped in water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 1 mL/min.
Length (Metric) 30 cm
Max. Pressure 20 bar
Particle Size 13 μm
pH 2.0 - 12
Pore Size 100 to 1000 Å
Solvents ≤20% organic solvent
Temperature 80°C
USP Type L38, L39
Stationary Phase Hydroxylated Methacrylate
Product Line TSKgel
Matrix Polymer Particle Platform
Phase Size Exclusion
Material Stainless Steel
Technique SEC HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.