Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ NH2-100 HPLC Column, 3 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022101 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
5 cm
15 cm
Diameter (Metric):
2 mm
4.6 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-102-2101 15 cm 4.6 mm
11-102-2098 5 cm 2 mm
11-102-2099 15 cm 2 mm
11-102-2100 5 cm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-102-2101 Supplier MilliporeSigma Supelco Supplier No. 821970
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel™ NH2-100 amino columns expand the range of TSKgel™ columns for hydrophilic interaction chromatography (HILIC). Offering a different selectivity from the well known TSKgel™ Amide-80 columns, these novel amino-bonded phase columns stand out by providing much improved chemical stability, a prerequisite for achieving reproducible and reliable results.

TSKgel™ NH2-100 columns are unique in that the bonded phase ligand not only, as expected, has a terminal primary amino group, but that the spacer also incorporates secondary as well as tertiary amino groups. Anionic compounds are retained on the column by ionic interaction. This allows for the use of gradients in salt concentration, in addition to gradient elutions with acetonitrile. Since the TSKgel™ NH2-100 columns have cationic sites, the columns can be used as mixed mode columns under some conditions. TSKgel™ NH2-100 columns are well suited for the analysis of all types of hydrophilic compounds, including carbohydrates and peptides. Due to a high ligand density and large surface area, these columns show stronger retention of polar compounds than TSKgel™ Amide-80 columns.

Order Info

Shipped in 85% acetonitrile - 15% water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 1 mL/min.
Length (Metric) 15 cm
Max. Pressure 150 bar
Particle Size 3 μm
pH 2.0 - 7.5
Pore Size 100 Å
Solvents 0 to 100% organic solvent
Surface Area 450 m2/g
Temperature 10°C to 50°C
USP Type L8
Stationary Phase Amine
Product Line TSKgel
Matrix Silica Particle Platform
Phase Normal Phase, HILIC, Ion Exchange
Material Stainless Steel Hardware
Technique Mixed Mode
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.