Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Octadecyl-2PW HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021905 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric):
2 mm
4.6 mm
6 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
11-102-1905 6 mm
11-102-1904 4.6 mm
11-102-1967 2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-1905 Supplier MilliporeSigma Supelco Supplier No. 817501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly cross-linked polymethacrylate base material provides excellent stability in high pH buffer systems. Withstands rigorous cleaning with either acid or base, 125 Å pores allow for analysis of peptides up to 8,000 Da.

  • Columns packed with larger particle sizes are available for semi-preparative scale separations
  • Polymer-based particles for reversed-phase (RP) HPLC are more pH tolerant, and prevent silanol interactions that are common with silica-based particles
  • Some silicas have an intrinsic metal content that may cause chelation problems with sensitive biomolecules, leading to poor recovery
  • Polymeric TSKgel Columns come in C18 and phenyl functional groups, as well as a range of particle porosities
  • Fast kinetics and quantitative protein recovery at submicrogram loading are characteristic of nonporous, resin-based TSKgel Octadecyl-NPR Columns
  • Shipped in 55% Acetonitrile, 45% Water
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 6 mm
Flow Rate 1.5 mL/min
Length (Metric) 15 cm
Ligand Type 1 meq/mL (Estimated Ligand Density)
Max. Pressure 120 bar
Particle Size 5 μm
pH 2 to 12
Pore Size 125 Å
Solvents 0-100% Organic Solvent
Temperature 4°C to 50°C
Stationary Phase C18 (octadecyl)
Product Line TSKgel
Shipping Condition Shipped in 55% Acetonitrile, 45% Water
Matrix C18 (octadecyl) Phase/Polymer Particle Platform
Phase Reversed Phase
Material Stainless Steel Hardware
Technique Reversed Phase HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.