Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Octadecyl-4PW HPLC Column, 7 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021968 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric):
2 mm
4.6 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
11-102-1968 2 mm
11-102-1838 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-1968 Supplier MilliporeSigma Supelco Supplier No. 818755
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides excellent stability in high pH buffer systems, can withstand rigorous cleaning with either acid or base.

The TSKgel Octadecyl-4PW HPLC column is a polymer-based, reversed-phase column designed for the analytical and preparative separation of water-soluble substances, including polysaccharides, nucleic acids, proteins, peptides, oligomers, and polymers.

  • Material: C18 (octadecyl) phase on polymer particle platform with stainless steel hardware
  • Pore Size: 500 Å (50 nm)
  • pH Range: 2 to 12
  • Temperature Range: 10°C to 80°C
  • Maximum Pressure: 100 bar (10.0 MPa)
  • Performance: Theoretical plates range from 2,000 to 5,000 per column; asymmetry factor 0.7
  • Compatibility: Suitable for aqueous solutions and water-soluble organic solvents (e.g., methanol, ethanol, acetonitrile, formic acid, DMSO)
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2 mm
Flow Rate 0.22 mL/min
Length (Metric) 15 cm
Ligand Type 1 meq/mL (Estimated Ligand Density)
Max. Pressure 100 bar
Particle Size 7 μm
pH 2 to 12
Pore Size 500 Å
Solvents 0-100% Organic Solvent
Temperature 5°C to 50°C
Stationary Phase C18 (octadecyl)
Product Line TSKgel
Shipping Condition Shipped in 60% Methanol, 40% Water
Matrix C18 (octadecyl) Phase/Polymer Particle Platform
Phase Reversed Phase
Material Stainless Steel Hardware
Technique Reversed Phase HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.