Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Octadecyl-NPR HPLC Column, 2.5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021842 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
3.5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1842 3.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1842 Supplier MilliporeSigma Supelco Supplier No. 814005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The highly cross-linked, polymethacrylate base material of TSKgel Octadecyl-NPR provides excellent stability in high pH buffer systems. It can withstand rigorous cleaning with either acid or base.

  • NPR, non-porous resin, columns are prepared from non-porous methacrylate particles of uniform 2.5 micron size
  • Provide high efficiency separations and fast analysis of peptides and proteins
  • The non-porous particle structure limits product isolation to sub-microgram loads
  • Polymer-based particles for reversed-phase (RP) HPLC are more tolerant of high and low pH operating conditions and eliminate silanol interactions
  • Some silicas have an intrinsic metal content that may cause chelation problems with sensitive biomolecules, leading to poor recovery
  • Polymeric TSKgel columns come in C18 and phenyl functional groups and a range of particle porosities
  • Fast kinetics and quantitative protein recovery at submicrogram loading are characteristic of nonporous, resin-based TSKgel Octadecyl-NPR Columns
  • Octadecyl-NPR particles are nonporous
  • Shipped in 70% Methanol, 30% Water
  • Salt Concentration: ≤ 0.5 M
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 1.6 mL/min
Length (Metric) 3.5 cm
Ligand Type 1 meq/mL (Estimated Ligand Density)
Max. Pressure 200 bar
Particle Size 2.5 μm
pH 2 to 12
Solvents 0-100% Organic Solvent
Temperature 0°C to 60°C
Stationary Phase C18 (octadecyl)
Product Line TSKgel
Shipping Condition Shipped in 70% Methanol, 30% Water
Matrix C18 (octadecyl) Phase/Polymer Particle Platform
Phase Reversed Phase
Material Stainless Steel
Technique Reversed Phase HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.