Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Octyl-80Ts HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021886 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
15 cm
25 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1886 25 cm
11-102-1885 15 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-1886 Supplier MilliporeSigma Supelco Supplier No. 817345
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel Octyl-80Ts has a lower carbon load and hydrophobicity than the corresponding ODS-80Ts products.

  • The C8 alkyl ligand provides a unique selectivity for the analysis of low MW pharmaceuticals, bases, nucleosides, and nucleotides
  • Features high purity, metal free silica and a proprietary technique for complete endcapping of residual silanol groups
  • Reduce tailing when analyzing basic compounds
  • Common applications include purity checks and peptide mapping
  • Shipped in 70% Methanol, 30% Water
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 1.2 mL/min
Length (Metric) 25 cm
Max. Pressure 300 bar
Particle Size 5 μm
pH 2 to 7.5
Pore Size 80 Å
Solvents 0-100% Organic Solvent
Surface Area 425 m2/g
Temperature 10°C to 50°C
USP Type L1, L7
Stationary Phase C8 (octyl)
Product Line TSKgel
Shipping Condition Shipped in 70% Methanol, 30% Water
Matrix C8 (octyl) Phase/Silica Particle Platform
Carbon Load 11%
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel Hardware
Technique Reversed Phase HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.