Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ ODS-100Z HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022156 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
1 cm
2 cm
3.5 cm
5 cm
7.5 cm
10 cm
15 cm
25 cm
Diameter (Metric):
2 mm
3 mm
4.6 mm
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-102-2156 1 cm 2 mm
11-102-2050 5 cm 2 mm
11-102-2157 2 cm 2 mm
11-102-2158 3.5 cm 2 mm
11-102-2051 15 cm 4.6 mm
11-102-2159 7.5 cm 2 mm
11-102-2052 25 cm 4.6 mm
11-102-2160 10 cm 2 mm
11-102-2161 25 cm 2 mm
11-102-2162 2 cm 3 mm
11-102-2163 3.5 cm 3 mm
11-102-2164 5 cm 3 mm
11-102-2165 7.5 cm 3 mm
11-102-2166 10 cm 3 mm
11-102-2167 15 cm 3 mm
11-102-2168 25 cm 3 mm
11-102-2169 2 cm 4.6 mm
11-102-2170 3.5 cm 4.6 mm
11-102-2171 5 cm 4.6 mm
11-102-2172 7.5 cm 4.6 mm
11-102-2173 10 cm 4.6 mm
11-102-2049 15 cm 2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Catalog No. 11-102-2156 Supplier MilliporeSigma Supelco Supplier No. 822748
Add to Cart
Add to Cart

TSKgel™ ODS-100Z column's bonded phase is prepared by reacting the silica surface with a difunctional octadecylsilane reagent, followed by repeated endcapping with a monofunctional trimethylsilane reagent.

TSKgel™ ODS-100Z columns contain a high density monomeric C18 bonded phase for maximum retention and selectivity of small molecular weight compounds and minimal secondary interaction with residual silanol groups. TSKgel™ ODS-100Z reversed phase columns are the workhorse for analysis of small molecular weight compounds in life science applications. Lot-to-lot reproducibility stands out.

Order Info

Shipped in acetonitrile.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2 mm
Flow Rate 0.2 mL/min.
Length (Metric) 1 cm
Max. Pressure 90 bar
Particle Size 5 μm
pH 2.0 - 7.5
Solvents 0 to 100% organic solvent
Surface Area 450 m2/g
Temperature 10°C to 50°C
USP Type L1
Stationary Phase C18 (octadecyl)
Product Line TSKgel
Matrix Spherical Silica
Carbon Load 20%
Phase Reversed Phase
Material Stainless Steel
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.