Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ TSKgel™ ODS-100Z HPLC Column phase C18 (octadecyl), 3 μm Particle Size, Stainless Steel

Catalog No. 111007422 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
7.5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-100-7422 7.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-7422 Supplier MilliporeSigma™ Supelco™ Supplier No. 822744
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This general purpose reversed phase column is the workhorse for analysis of small molecular weight compounds in life science applications.

The bonded phase of TSKgel™ ODS-100Z columns is prepared by reacting the silica surface with a difunctional octadecylsilane reagent, followed by repeated endcapping with a monofunctional trimethylsilane reagent. The resulting TSKgel™ ODS-100Z columns contain a high density monomeric C18 bonded phase for Maximum retention and selectivity of small molecular weight compounds and minimal secondary interaction with residual silanol groups.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 7.5 cm
Particle Size 3 μm
pH 2.0 to 7.5
Surface Area 450 m2/g
Stationary Phase C18 (octadecyl)
Product Line TSKgel
Matrix C18 (octadecyl) Phase
Carbon Load 20%
Phase Reversed Phase
Material Stainless Steel
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.