Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ ODS-120A HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021762 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric):
7.8 mm
21.5 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
11-102-1762 7.8 mm
11-102-1752 21.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-1762 Supplier MilliporeSigma Supelco Supplier No. 807129
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel™ ODS-120A columns contain 150 Å pore size silica particles. The exclusion limit of 10,000 Da makes TSKgel™ ODS-120A columns a good choice for the reversed phase chromatography of peptides, small proteins, and environmental samples such as poly-aromatic hydrocarbons.

The 'A' in the product description signifies that the material is not endcapped. For charged samples, the endcapped TSKgel™ ODS-120T column is a more suitable alternative.

Order Info

Shipped in 70% methanol - 30% water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 3 mL/min.
Length (Metric) 30 cm
Max. Pressure 75 bar
Particle Size 10 μm
pH 2.0 - 7.5
Pore Size 150 Å
Solvents 0 to 100% organic solvent
Temperature 0°C to 45°C
USP Type L1
Stationary Phase C18 (octadecyl)
Product Line TSKgel
Matrix Spherical Silica
Carbon Load 20%
Endcapped No
Phase Reversed Phase
Material Stainless Steel
Technique Reversed Phase HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.