Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ ODS-120T HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021764 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric):
7.8 mm
21.5 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
11-102-1764 21.5 mm
11-102-1763 7.8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-1764 Supplier MilliporeSigma Supelco Supplier No. 807134
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

TSKgel™ ODS-120T columns contain 150 Å pore size silica particles. The exclusion limit of 10,000 Da makes TSKgel™ ODS-120T columns a good choice for the reversed phase chromatography of peptides, small proteins, and small molecular weight compounds in organic and environmental samples.

The columns are endcapped with trimethyl silane groups to improve the peak shape of positively charged analytes.

Order Info

Shipped in 70% methanol - 30% water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 21.5 mm
Flow Rate 12 mL/min.
Length (Metric) 30 cm
Max. Pressure 60 bar
Particle Size 10 μm
pH 2.0 - 7.5
Pore Size 150 Å
Solvents 0 to 100% organic solvent
Temperature 10°C to 50°C
USP Type L1
Stationary Phase C18 (octadecyl)
Product Line TSKgel
Matrix Spherical Silica
Carbon Load 22%
Phase Reversed Phase
Material Stainless Steel
Technique Reversed Phase HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.