Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ ODS-80Tm HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021805 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
15 cm
25 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1805 15 cm
11-102-1806 25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-1805 Supplier MilliporeSigma Supelco Supplier No. 808148
Add to Cart
Add to Cart

TSKgel™ ODS-80Tm is a general purpose column for the analysis of low MW pharmaceuticals, bases, nucleosides and nucleotides.

Common applications include purity checks and peptide mapping. Bonded phase pore size is indicated by the number in the product description, in this case TSKgel™ ODS-80Tm has 80 Å nominal pore size. The pore size of the base silica is 100 Å.

Order Info

Shipped in 70% methanol - 30% water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 1.2 mL/min.
Length (Metric) 15 cm
Max. Pressure 210 bar
Particle Size 5 μm
pH 2.0 - 7.5
Pore Size 100 Å
Solvents 0 to 100% organic solvent
Surface Area 425 m2/g
Temperature 0°C to 45°C
USP Type L1
Stationary Phase C18 (octadecyl)
Product Line TSKgel
Matrix Silica Particle Platform
Carbon Load 15%
Endcapped Yes, Bifunctional Dialkylsilane
Phase Reversed Phase
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.