Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Phenyl-5PW RP HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021843 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Glass
Stainless Steel Hardware
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
11-102-1843 Glass
11-102-1969 Stainless Steel Hardware
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-1843 Supplier MilliporeSigma Supelco Supplier No. 814007
Add to Cart
Add to Cart

TSKgel Phenyl-5PW RP offers similiar selectivity to its hydrophobic interation counterpart TSKgel Phenyl-5PW, although a higher degree of cross-linking allows for increased stability in aqueous/organic mobile phase gradients. Provides an advantage over silica when high pH buffer systems are needed.

  • TSKgel Phenyl-5PW RP can withstand rigorous cleaning protocols using either acid or base
  • The 1000 Å pore size accommodates globular protein samples up to 1,000,000 Da.
  • Polymer-based particles for reversed-phase (RP) HPLC are more tolerant of high and low pH operating conditions
  • They do not have silanol interactions that may cause peak tailing in some basic analytes
  • Some silicas have an intrinsic metal content that may cause chelation problems with sensitive biomolecules, leading to poor recovery
  • Shipped in 30% methanol, 70% water
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 8 mm
Flow Rate 2.5 mL/min
Length (Metric) 7.5 cm
Ligand Type 1 meq/mL (Estimated Ligand Density)
Max. Pressure 20 bar
Particle Size 10 μm
pH 2 to 12
Pore Size 1000 Å
Solvents 0-100% Organic Solvent
Temperature 4°C to 60°C
Stationary Phase Phenyl (octadecyl)
Product Line TSKgel
Shipping Condition Shipped in 30% Methanol, 70% Water
Matrix Phenyl (octadecyl) Phase/Hydroxylated Methacrylic Polymer
Phase Reversed Phase
Material Glass
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.