Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Phenyl-5PW RP HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021803 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
7.5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1803 7.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1803 Supplier MilliporeSigma Supelco Supplier No. 808043
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel™ Phenyl-5PW RP columns are filled with 10 micron porous polymethacrylate particles, the surface of which is covered with a high density of phenyl groups that causes proteins to be retained in a low salt buffer mobile phase and requiring a solvent gradient for their elution, as is the protocol in reversed phase chromatography.

In contrast, TSKgel™ Phenyl-5PW has such a low density of phenyl groups on the surface that proteins can only be retained at high salt concentration and eluted by a decreasing salt gradient, which, of course, is the protocol used in hydrophobic interaction chromatography. TSKgel™ Phenyl-5PW RP columns can withstand rigorous cleaning with either acid or base. The 1000 Å pore size accommodates globular protein samples up to 1,000,000 Da.

Order Info

Shipped in 30% methanol - 70% water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 1.2 mL/min.
Length (Metric) 7.5 cm
Ligand Type 1 meq/mL ligand density (estimate)
Max. Pressure 30 bar
Particle Size 10 μm
pH 2.0 - 12
Pore Size 1000 Å
Solvents 0 to 100% organic solvent
Temperature 4°C to 60°C
Stationary Phase Phenyl
Product Line TSKgel
Matrix Polymer Particle Platform
Phase Reversed Phase
Material Stainless Steel
Technique Reverse Phase HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.