Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Protein C4-300 HPLC Column, 3 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. p-7213973 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
2 cm
5 cm
10 cm
15 cm
Diameter (Metric):
2 mm
4.6 mm
5 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-102-2187 15 cm 2 mm
11-102-2182 5 cm 4.6 mm
11-102-2183 10 cm 4.6 mm
11-102-2184 15 cm 4.6 mm
11-102-2185 2 cm 5 mm
11-102-2186 10 cm 2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 11-102-2187 Supplier MilliporeSigma Supelco Supplier No. 822832
Only null left
Add to Cart
Add to Cart

TSKgel™ Protein C4-300 high efficiency columns are designed for the optimal recovery and resolution of proteins such as recombinant proteins, antibody fragments or PEGylated proteins.

TSKgel™ Protein C4-300 column's utilize the latest surface modification techniques and endcapping of residual silanol groups reduce undesirable secondary interactions and peak tailing.

Order Info

Shipped in acetonitrile.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2 mm
Flow Rate 1 mL/min.
Length (Metric) 15 cm
Max. Pressure 300 bar
Particle Size 3 μm
pH 1.5 - 7.5
Solvents 0 to 100% organic solvent
Temperature 10°C to 50°C
USP Type L26
Stationary Phase C4 (butyl)
Product Line TSKgel
Matrix Spherical Silica
Carbon Load 3%
Endcapped Yes, Trimethylsilane
Phase Reversed Phase
Material Stainless Steel
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.