Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ QAE-2SW HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021771 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
25 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1771 25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1771 Supplier MilliporeSigma Supelco Supplier No. 807166
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel™ QAE-2SW columns are packed with 5 μm porous spherical silica beads which are surface modified with a strong anion exchanger. They are used for the separation and analysis of peptides, nucleotides, nucleosides and other biologically active molecules.

TSKgel® QAE-2SW columns are designed for the separation and analysis of peptides, nucleotides, nucleosides, and other biologically active molecules via anion exchange chromatography.

  • Matrix: Porous spherical silica
  • Stationary Phase: Quaternary ammonium ion
  • Particle Size: 5 μm
  • Column Dimensions: 25 cm x 4.6 mm
  • USP Type: L14
  • Operating pH: 2.0-7.5
  • Temperature Range: 10°C to 45°C
  • Max Pressure: 150 bar
  • Solvent Compatibility: 0-100% organic solvent
  • Flow Rate: 1 mL/min
  • Molecular Weight Range: 100-5000
  • Construction Material: Stainless steel

Order Info

Shipped in methanol.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 1 mL/min.
Length (Metric) 25 cm
Max. Pressure 150 bar
Particle Size 5 μm
pH 2.0 - 7.5
Solvents 0 to 100% organic solvent
Temperature 10°C to 45°C
USP Type L14
Stationary Phase Ammonium ion, quaternary
Product Line TSKgel
Matrix Spherical Silica
Phase Anion Exchange
Material Stainless Steel
Technique Anion Exchange HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.