Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ SP-5PW HPLC Column, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021971 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric):
2 mm
5 mm
8 mm
20 mm
Material:
Glass
Stainless Steel Hardware
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Material
11-102-1971 2 mm Stainless Steel Hardware
11-102-1828 5 mm Glass
11-102-1822 8 mm Glass
11-102-1847 20 mm Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-102-1971 Supplier MilliporeSigma Supelco Supplier No. 818758
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For the separation and analysis of proteins, peptides, and other biologically active molecules.

  • Packed with porous hydrophilic polymer beads that are surface modified with a strong cation exchanger
  • For the separation and analysis of proteins, peptides, and other biologically active molecules
  • Columns have the same backbone chemistry and selectivity as the bulk process scale resins Toyopearl SP-650 & TSKgel SP-5PW (20) and (30)
  • Salt Concentration: ≤1 M
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2 mm
Flow Rate 0.12 mL/min
Ion Exchange Group Na+
Length (Metric) 7.5 cm
Max. Pressure 10 bar
Particle Size 10 μm
pH 2 to 12
Pore Size 1000 Å
Solvents ≤ 20% Organic Solvent
Temperature 10°C to 45°C
Stationary Phase Sulfonic Acid
Product Line TSKgel
Matrix Sulfonic Acid Phase/Hydroxylated Methacrylic Polymer
Phase Cation Exchange
Material Stainless Steel Hardware
Technique Cation Exchange
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.