Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ SP-NPR HPLC Column, 2.5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021835 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
3.5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1835 3.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1835 Supplier MilliporeSigma Supelco Supplier No. 813076
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel SP-NPR columns are packed with non-porous, hydrophilic polymer beads which are surface modified with a strong cation exchanger. They are used for the separation and analysis of proteins and peptides.

  • Particularly useful for adeno-associated viruses and other large macromolecules
  • Salt Concentration: ≤ 1 M
  • Shipped in DI water
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 1.6 mL/min
Ion Exchange Group Na+
Length (Metric) 3.5 cm
Max. Pressure 200 bar
Particle Size 2.5 μm
pH 2 to 12
Pore Size Nonporous
Solvents ≤ 20% Organic Solvent
Temperature 10°C to 60°C
Stationary Phase Sulfonic Acid
Product Line TSKgel
Shipping Condition Shipped in DI Water
Matrix Sulfonic Acid Phase/Polymer Particle Platform
Phase Strong Cation Exchange
Material PEEK
Technique Cation Exchange
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.