Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ SP-STAT HPLC Column, 7 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022095 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
10 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-2095 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2095 Supplier MilliporeSigma Supelco Supplier No. 821964
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel™™ SP-STAT columns are packed with hydrophilic non-porous resin particles of which the surface consists of an open access network of multi-layered cation exchange groups.

TSKgel™™ SP-STAT's innovative bonding chemistry, combined with a relatively large particle size, results in a respectable loading capacity and a low operating pressure. Applications for the TSKgel™ SP-STAT columns include the separation of proteins, protein aggregates, charge isomers of monoclonal antibodies, PEGylated proteins, and peptide digests.

Order Info

Shipped in DI water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 0.5 to 1.4 mL/min.
Length (Metric) 10 cm
Max. Pressure 100 bar
Particle Size 7 μm
pH 3.0 - 10
Solvents ≤50% organic solvent
Temperature 10°C to 60°C
Stationary Phase Sulfonic Acid
Product Line TSKgel
Matrix Polymer Particle Platform
Phase Cation Exchange
Material Stainless Steel
Technique Cation Exchange
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.