Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Super-Octyl HPLC Column, 2.3 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022035 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
5 cm
10 cm
Diameter (Metric):
2 mm
4.6 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-102-2035 10 cm 2 mm
11-102-2034 5 cm 2 mm
11-102-1936 10 cm 4.6 mm
11-102-1935 5 cm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-102-2035 Supplier MilliporeSigma Supelco Supplier No. 820014
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel™ Super-Octyl Columns are packed with monodispersed, 2 μm (nominal, mean particle size is 2.3 μm) spherical silica particles covalently bonded with octyl groups.

  • A small particle size makes the Super series the highest efficiency, reversed phase columns in the TSKgel product line
  • Monodispersed packing generates operational back pressures more typical of larger particles, allowing the use of higher flow rates than other 2 μm packings
  • Offers less hydrophobicity than TSKgel Super-ODS
  • An excellent choice for peptides, proteins, amino acids, tryptic digests, nucleotides, pharmaceutical molecules, and food/beverage samples
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2 mm
Flow Rate 0.25 mL/min
Length (Metric) 10 cm
Max. Pressure 300 bar
Particle Shape Sphere
Particle Size 2.3 μm
pH 2 to 7.5
Solvents 0-100% Organic Solvent
Surface Area 100 m2/g
Temperature 10°C to 50°C
USP Type L7
Stationary Phase C8 (octyl)
Product Line TSKgel
Matrix C8 (octyl) Phase/Silica Particle Platform
Carbon Load 5%
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Technique Reversed Phase HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.