Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ SuperH2500 HPLC Column, 3 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021910 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
15 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1910 15 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1910 Supplier MilliporeSigma Supelco Supplier No. 817992
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Packed with rigid, porous polymer beads and used for the non-aqueous separation of polymers with molecular weights less than 20,000 Da.

  • A high efficiency/high throughput version of the conventional TSKgel HHR Column
  • Both column types are based on the same bead chemistry
  • High-throughput gel permeation columns have dimensions of 6 mm ID x 15 cm
  • Shipped in THF, which can be exchanged for a wide variety of organic solvents
  • Maximum Operating Temperature: 140°C
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 6 mm
Flow Rate 0.8 mL/min
Length (Metric) 15 cm
Max. Pressure 60 bar
Particle Size 3 μm
Pore Size 30 Å
Temperature 140°C
USP Type L21
Stationary Phase Polystyrene-divinylbenzene
Product Line TSKgel
Matrix Polystyrene-divinylbenzene Phase/Polymer Particle Platform
Material Stainless Steel Hardware
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.